  • Search Results
  • Exposure to first line anti-tuberculosis drugs in prepubertal age reduces the quality and functional competence of spermatozoa and oocytes in Swiss albino mice.
Exposure to first line anti-tuberculosis drugs in prepubertal age reduces the quality and functional competence of spermatozoa and oocytes in Swiss albino mice.
Environmental toxicology and pharmacology (2019-11-26)
Arpitha Rao, Guruprasad Nayak, Sandhya Kumari, Sneha Guruprasad Kalthur, Sadhana P Mutalik, Srinivas Mutalik, Satish Kumar Adiga, Guruprasad Kalthur
ABSTRACT
Prepubertal Swiss albino mice of both sex were administered with first-line anti-tuberculosis drugs (ATDs) viz; rifampicin, isoniazid, pyrazinamide, streptomycin and ethambutol intraperitoneally, for 4 weeks. Two weeks after the completion of treatment, male mice were sacrificed to collect caudal spermatozoa and female mice were superovulated with pregnant mare serum gonadotropin (PMSG) and human chorionic gonadotropin (hCG) to collect metaphase II (MII) oocytes from oviduct. Administration of ATDs not only decreased the count, motility and, nuclear maturity and also, increased the head abnormalities, mitochondrial damage and DNA damage in epididymal spermatozoa. Reduction in number of ovulated oocytes, increased degeneration rate and altered distribution pattern of cytoplasmic organelles was observed in oocytes of female mice. Presence of ATDs in in vitro maturation (IVM) medium increased abnormalities in meiotic resulted in abnormal spindle organization (except ethambutol) without affecting nuclear maturation. In conclusion, the result of this study indicates that ATDs have considerable adverse effects on the functional competence of male and female gametes, however, with varied degree of toxicity.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Medium 199, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Fluoroshield with PI, histology mounting medium
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Chromomycin A3 from Streptomyces griseus, ≥95% (HPLC)