  • Search Results
  • SET protein modulates H4 histone methylation status and regulates miR-137 level in oral squamous cell carcinoma.
SET protein modulates H4 histone methylation status and regulates miR-137 level in oral squamous cell carcinoma.
Epigenomics (2020-04-09)
Lucas Oliveira Sousa, Lays Martin Sobral, Luciana Oliveira de Almeida, Cristiana Bernadelli Garcia, Lewis Joel Greene, Andréia Machado Leopoldino
ABSTRACT
Aim: Histone acetylation and methylation control gene expression. We investigated the impact of SET knockdown on histone methylation status and the consequences for the miRNAs levels in oral squamous cell carcinoma (OSCC). Methods: OSCC cells with and without SET knockdown were analyzed by quantitative real-time PCR to determine miRNA levels, and by immunoreactions to histone modifications. Results: The knockdown of SET increased the levels of histone H4K20me2 and miR-137. Still, SET protein binds to the miR-137 promoter region. The transfection of miR-137 mimic reduced the KI67 and Rb proteins and proliferation of OSCC cells. Conclusion: Our results show for the first time a relationship between SET and histone methylation associated with the control of miRNA expression and KI67 and Rb as targets of miR-137 in OSCC.
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Goat serum
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
DAPI, for nucleic acid staining