  • Search Results
  • Cytosolic Ca2+ measurements by ratiometric fluorescence microscopy in melanoma cells.
Cytosolic Ca2+ measurements by ratiometric fluorescence microscopy in melanoma cells.
STAR protocols (2021-02-04)
Tiago Rodrigues, Letícia Silva Ferraz
ABSTRACT
Cytosolic Ca2+ levels are maintained at low nanomolar concentrations, and disruption of Ca2+ homeostasis is associated with cell/tissue damage. Thus, methods have been developed to accurately assess cellular Ca2+ levels, each with intrinsic advantages and disadvantages. Here, we present in detail a ratiometric fluorometric method for cytosolic Ca2+ measurement in cultured melanoma cells using Fura 2-AM cell loading and fluorescence microscopy imaging. For complete details on the use and execution of this protocol, please refer to Esteves et al. (2020).
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture