Merck
  • Al and Zn complexes bearing N,N,N-tridentate quinolinyl anilido-imine ligands: synthesis, characterization and catalysis in L-lactide polymerization.
Al and Zn complexes bearing N,N,N-tridentate quinolinyl anilido-imine ligands: synthesis, characterization and catalysis in L-lactide polymerization.
Dalton transactions (Cambridge, England : 2003) (2012-08-16)
Nan Yang, Lan Xin, Wei Gao, Jingshun Zhang, Xuyang Luo, Xiaoming Liu, Ying Mu
ABSTRACT
Reactions of N,N,N-tridentate quinolinyl anilido-imine ligands with AlMe(3) afford mononuclear aluminum complexes {κ(3)-[{2-[ArN=C(H)]C(6)H(4)}N(8-C(9)H(6)N)]}AlMe(2) (Ar = 2,6-Me(2)C(6)H(3) (1a), 2,6-Et(2)C(6)H(3) (1b), 2,6-(i)Pr(2)C(6)H(3) (1c)) or dinuclear complexes AlMe(3){κ(1)-[{2-[ArN=C(H)C(6)H(4)]N(8-C(9)H(6)N)}-κ(2)]AlMe(2) (R = 2,6-Me(2)C(6)H(3) (2a), 2,6-Et(2)C(6)H(3) (2b), 2,6-(i)Pr(2)C(6)H(3) (2c)) depending on the ratios of reactants used. Similar reactions of ZnEt(2) with these ligands give the monoligated ethyl zinc complexes {κ(3)-[{2-[ArN=C(H)]C(6)H(4)}N(8-C(9)H(6)N)]}ZnEt (Ar = 2,6-Me(2)C(6)H(3) (3a), 2,6-Et(2)C(6)H(3) (3b), 2,6-(i)Pr(2)C(6)H(3) (3c)) or bisligated complexes {κ(3)-[{2-[ArN=C(H)]C(6)H(4)}N(8-C(9)H(6)N)]}Zn{κ(2)-[{2-[ArN=C(H)]C(6)H(4)}N(8-C(9)H(6)N)]} (Ar = 2,6-Me(2)C(6)H(3) (4a), 2,6-Et(2)C(6)H(3) (4b), 2,6-(i)Pr(2)C(6)H(3) (4c)). These complexes were well characterized by NMR and the structures of 1a, 2a, 2c, 3b and 4c were confirmed by X-ray diffraction analysis. The aluminum and zinc complexes were tested to initiate lactide polymerization in which the zinc complexes show moderate to high activities in the presence of benzyl alcohol.
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Sigma-Aldrich
Quinoline, reagent grade, 96%
Sigma-Aldrich
Quinoline, reagent grade, 98%