Merck
  • Synthesis of poly (arylene ether amide) s containing CF 3 groups by nitro displacement reaction of AB-type monomers.
Synthesis of poly (arylene ether amide) s containing CF 3 groups by nitro displacement reaction of AB-type monomers.
Macromolecular Rapid Communications (2002)
Lee HS and Kim SY.