  • Search Results
  • Morphology and ionic conductivity of complexes of sodium iodide and sodium thiocyanate with poly (ethylene oxide).
Morphology and ionic conductivity of complexes of sodium iodide and sodium thiocyanate with poly (ethylene oxide).
Polymer (1982)
Lee CC and Wright PV.