  • Search Results
  • Valve interstitial cell culture: Production of mature type I collagen and precise detection.
Valve interstitial cell culture: Production of mature type I collagen and precise detection.
Microscopy research and technique (2017-04-30)
Jana Liskova, Daniel Hadraba, Elena Filova, Miroslav Konarik, Jan Pirk, Karel Jelen, Lucie Bacakova
ABSTRACT
Collagen often acts as an extracellular and intracellular marker for in vitro experiments, and its quality defines tissue constructs. To validate collagen detection techniques, cardiac valve interstitial cells were isolated from pigs and cultured under two different conditions; with and without ascorbic acid. The culture with ascorbic acid reached higher cell growth and collagen deposition, although the expression levels of collagen gene stayed similar to the culture without ascorbic acid. The fluorescent microscopy was positive for collagen fibers in both the cultures. Visualization of only extracellular collagen returned a higher correlation coefficient when comparing the immunolabeling and second harmonic generation microscopy images in the culture with ascorbic acid. Lastly, it was proved that the hydroxyproline strongly contributes to the second-order susceptibility tensor of collagen molecules, and therefore the second harmonic generation signal is impaired in the culture without ascorbic acid.
Sigma-Aldrich
Monoclonal Anti-Myosin (Smooth) antibody produced in mouse, clone hSM-V, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Calponin antibody produced in mouse, clone hCP, ascites fluid
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid