Merck
HPLC Analysis of Various Cationic Polymers on TSKgel® PWXL-CP Columns
HPLC Analysis of Various Cationic Polymers on TSKgel® PWXL-CP Columns application for HPLC
Materials
related product
Product No.
Description
Pricing
Ultra-low Range Molecular Weight Marker (M.W. 1,060-26,600)
Kit for Molecular Weights 14,000-500,000 Non-denaturing
TSKgel® G3000PWXL-CP HPLC Column
phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size
TSKgel® G5000PWXL-CP HPLC Column
phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 10 μm particle size
TSKgel® G6000PWXL-CP HPLC Column
phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size
used together
Product No.
Description
Pricing
Sodium nitrate
99.995% trace metals basis
TSKgel® PWXL-CP Guard Column
phase hydroxylated methacrylate, L × I.D. 4 cm × 6 mm, 13 μm particle size
CONDITIONS
column
TSKgel G3000PWXL-CP, 30 cm x 7.8 mm, 7 μm particles (821873)
column
TSKgel G5000PWXL-CP, 30 cm x 7.8 mm, 10 μm particles (821874)
column
TSKgel G6000PWXL-CP, 30 cm x 7.8 mm, 13 μm particles (821875)
mobile phase
0.1 M sodium nitrate
flow rate
1 mL/min
column temp.
25 °C
detector
refractive index (RI)
injection
100 μL
sample
3 mg/mL
Description
Analysis Note
Various cationic polymers with different functional groups and molecular weights were injected on the TSKgel PWXL-CP columns (TSKgel G6000PWXL-CP, G5000PWXL-CP and G3000PWXL-CP, connected in series).
Other Notes
Data provided by Tosoh Bioscience LLC.
Legal Information
TSKgel is a registered trademark of Tosoh Corporation