Merck
  • A1847
All Photos(1)

A1847

Sigma-Aldrich

[Asn670, Sta671, Val672]-Amyloid β/A4 Precursor Protein 770 Fragment 662-675 ammonium salt

≥95% (HPLC)

Empirical Formula (Hill Notation):
C73H118N16O27
Molecular Weight:
1651.81
MDL number:

assay

≥95% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Lys-Thr-Glu-Glu-Ile-Ser-Glu-Val-Asn-Sta-Val-Ala-Glu-Phe

General description

Potent substrate analog inhibitor of β-Secretase.

Packaging

1 mg

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service