Merck
All Photos(1)

43623

Supelco

β-Sitosterol

analytical standard

Synonym(s):
α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

mp

136-140 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

β-Sitosterol is an abundantly found phytosterol in plants. It is known to show anti-inflammatory, immune-modulating, antipyretic, antineoplastic, and blood sugar-controlling properties. β-Sitosterol also acts as an estrogen and estrogen receptor (ERα and Erβ) agonist.

Application

This analytical standard can also be used as follows:
  • Identification and determination of β-sitosterol and stigmasterol in the hydro-alcohol extract, prepared from the leaves of B. bambo by reversed-phase high-performance liquid chromatography (RP-HPLC) coupled to photo-diode array detector (PDA)
  • Development and validation of two high-performance thin layer chromatography (HPTLC) methods to quantify four biomarkers— β-sitosterol, β-amyrin, lupeol, and ursolic acid in the ethanol extract of G. senegalensis leaves
  • Secondary metabolite profiling of various plant parts collected from 82 plants belonging to 21 different cannabis strains using gas chromatography-mass spectrometry (GC-MS) for sterols and terpenoids (mono-, sesqui-, tri-), and high-performance liquid chromatography (HPLC) with UV and mass spectrometric (MS) detection for flavonoids
  • High-performance thin layer chromatographic (HPTLC) method-based quantification of β-sitosterol in a herbal formulation prepared from the roots of Gmelina arborea using silica gel precoated 60 F254 aluminum plates
  • Determination of free sterols or oxysterols in food matrices, using gas chromatography and ion trap mass spectrometry (GC-MS).

Packaging

10 mg
Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service