Merck
All Photos(1)

24610C

SAFC

EX-CELL® 610-HSF Serum-Free Medium for Hybridoma Cells

Low-protein, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture

MDL number:
NACRES:
NA.71

Quality Level

description

for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
14621C14610C24621C
form

dry powder

form

liquid

form

liquid

form

dry powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

400

Quality Level

400

Quality Level

500

description

for research or for further manufacturing use

description

for research or for further manufacturing use

description

Drug Master File available, for research or for further manufacturing use

description

for research or for further manufacturing use

Application

EX-CELL 610-HSF is a low-protein (11 mg/L) serum-free medium. Originally developed to support the growth of hybrid cells in culture, it has been shown to support a wide range of cells including lymphoid and epithelial cells and B cell hybridomas of murine, rat and human origin. EX-CELL 610-HSF has been used in stationary culture systems and in large-scale bioreactors. In both types of culture, the production of cellular products, particularly monoclonal antibodies, has been shown to equal or exceed levels seen when the same cells are cultured in the presence of Fetal Bovine Serum (FBS).

Quantity

Formulated to contain 15.1 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service