Merck
All Photos(1)

66540369

Sigma-Aldrich

BIONi010-C-7

Human iPS Cell Line

Synonym(s):
EBiSC iPSC Line, Human iPSC, Induced Pluripotent Stem Cell Line, iPS Cell, iPSC

biological source

human dermis (fibroblast)

reprogramming method

episomal

description

age (15-19)

manufacturer/tradename

EBiSC

gender

male

growth mode

adherent (pluripotent)

technique(s)

cell culture | stem cell: suitable

relevant disease(s)

Alzheimer′s disease

shipped in

dry ice

storage temp.

−196°C

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Cell Line Origin

Depositor
Bioneer

Cell Line Description

Derivation
Primary cell type: Fibroblast of dermis
Primary cell developmental stage: Adult
Reprogramming method
Vector type: Non-integrating
Vector: Episomal
Gene list:
KLF4
Lin28
MYC
POU5F1
shP53
SOX2
Other:From the same donor: BIONi010-A, BIONi010-B

Characterization
Differentiation potency
Ectoderm: HES5(+),NeuroD1(+),PAX6 (+),Endoderm: CXCR4(+),FOXA2 (+),GATA6 (+),GSC (+)PITX1(+)SOX17 (+),Mesoderm: DCN(+)NCAM1 (+)VIM(+)

Microbiology / Virus Screening
HIV 1: Negative
HIV 2:Negative
Hepatitis B: Negative
Hepatitis C:Negative
Mycoplasma:Negative
Sterility
Inoculation for microbiological growth: No Contaminants Detected
Mycoplasma: Not Detected
Viability: Viable post-cryopreservation

Genotyping
Karyotyping
Passage number: 27
Cell line karyotype: 46 XY
Karyotyping method: G-Banding
Genotyping
STR/Fingerprinting: A 16 allele profile has been recorded and data is available upon request, after cell line purchase.

Genetic Modification
Disease: Alzheimer′s disease
Type of modification: Isogenic
Gene: TREM2
Chromosome location: 6p21.1
Nucleotide sequence HGSV: rs75932628(T)
Protein sequence HGSV: R47H
Zygosity: Homozygous
Target locus modification: Mutated

Linkage

Note: EAUA and CLIP must be completed before order fulfillment

Subculture Routine

Medium: Essential E8®
Passage method: EDTA
Matrix: Matrigel® / Geltrex®
CO2 concentration: 5%
O2 concentration: 18%
Temperature: 37°C

Legal Information

E8 is a registered trademark of WIsconsin Alumni Research Foundation non-stock Corporation
EBiSC is a trademark of Fraunhofer-Gesellschaft
GELTREX is a registered trademark of Life Technologies Corporation
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service