Merck
All Photos(2)

CN492-05

Sigma-Aldrich

Canine Marrow Stromal Cells: CnMSC (Cryovial)

NACRES:
NA.81

biological source

canine bone marrow (normal)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 78

morphology

stromal

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

tuberculosis

shipped in

dry ice

storage temp.

−196°C

General description

Bone marrow is the major blood creating organ, but in addition to supporting hematopoietic growth and differentiation, marrow stromal cells can be induced to produce cells of other connective tissues, such as bone, cartilage, and fat, as well as cells from neuroectodermal (neurons) and endodermal (hepatocytes) lineages. The potential of HMSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.

Cell Line Origin

Bone Marrow

Application

blood production, hematopoietic growth and differentiation, pathways of pluripotent cell differentiation into bone, cartilage and fat, connective tissue production,supportive structure, cell production, cell proliferation, cell based therapy, tissue restoration and repair, differentiation, effects of culture conditinos, HLA expression, cytokine production, immunomodulation, clinical trials

Components

Canine Marrow Stromal Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Canine Marrow Stromal Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the CnMSC Culture Protocol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service