Merck
All Photos(2)

RAB0306

Sigma-Aldrich

Human IL-6 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):
B-cell stimulatory factor 2, CTL differentiation factor, Hybridoma growth factor, IFN-beta-2, IL-6, Interferon beta-2, Interleukin-6
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 1.37-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL6(3569)

General description

The Human IL-6 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-6 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human I-L6

Application

Human IL-6 ELISA kit has been used for determining the levels of IL-6 in biological samples. [1][2][3]
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

IL-6 (interleukin 6) is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism.[4] IL-6 is associated with various autoimmune and inflammatory diseases. High levels of IL-6 are present in the synovial fluid from rheumatoid arthritis patients. It is also linked with chronic inflammatory proliferative disease and B cell malignancy, Castleman′s disease, and plasmacytoma/multiple myeloma.[4]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)

  • RABSTOP3ELISA Stop Solution (Item I)

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)

  • RABWASH420X Wash Buffer (Item B)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service