Merck
All Photos(2)

55314-U

Supelco

Supel Tox AflaZea

volume 6 mL, pk of 30

Synonym(s):
Supel Tox
NACRES:
NB.21

material

polypropylene tube

Quality Level

product line

Supel

composition

bed wt., 1.2 g

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

matrix active group

Proprietary (Mycotoxins) phase

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

The need for a quick, simplistic sample cleanup approach prior to chromatographic mycotoxin analysis has brought about SPE cartridges that significantly decrease sample prep time, increase reproducibility, and are more user friendly as compared to the industry standard immunoaffinity columns. In addition the Supel Tox SPE approach requires less equipment and fewer consumables, providing an additional cost savings .

Unlike the multiple step “bind and elute” strategy implemented when using immunoaffinity columns, the Supel Tox AflaZea, DON, and Tricho SPE cartridges employ an “interference removal” strategy which saves time by eliminating wash steps prior to elution of aflatoxin, zearalenone, deoxynivalenol, and tricothecenes (type A and B), respectively. Cartridges removing interferences associated with analysis of the above and fumonisins (B1 and B2) as well as ochratoxin A are also available as a part of our Supel Tox product offering.

Features and Benefits

  • Removes interferences associated with mycotoxin analysis.
  • Better reproducibility than the current industry standard, immunoaffinity columns.
  • Time associated with sample preparation is up to ten times faster than that related to immunoaffinity columns.
  • Basic and quick methodology requiring little additional method development.
  • Improved shelf life over immunoaffinity columns due to thermally stable format. No refrigeration required for shipping and storage.

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service