  • Search Results
  • Effect of the absence or presence of various protein supplements on further development of bovine oocytes during in vitro maturation
Effect of the absence or presence of various protein supplements on further development of bovine oocytes during in vitro maturation
Biology of Reproduction (2002)
Ali A and Sirard M