HK
EN
Applications
Products
Services
Support
Home
Search Results
Identification of a Golgi-localised GRIP domain protein from Arabidopsis thaliana
Identification of a Golgi-localised GRIP domain protein from Arabidopsis thaliana
Planta (2004)
Gilson PR, et al.