HK
EN
Applications
Products
Services
Support
Home
Search Results
Isolation and culture of human endometrial epithelial cells and stromal fibroblasts
Isolation and culture of human endometrial epithelial cells and stromal fibroblasts
Bio-protocol (2015)
Chen JC and Roan NR