Merck
  • Keratin 8 phosphorylation in vitro by cAMP-dependent protein kinase occurs within the amino- and carboxyl-terminal end domains.
Keratin 8 phosphorylation in vitro by cAMP-dependent protein kinase occurs within the amino- and carboxyl-terminal end domains.
Biochemical and biophysical research communications (1996-04-05)
S Ando, T Tokui, T Yano, M Inagaki
ABSTRACT
We reported earlier that phosphorylation in vitro of keratin filaments reconstituted from rat type I keratin 18 and type II keratin 8 by cAPM-dependent protein kinase induces disassembly of the keratin filament structure. Keratin 8 rather than keratin 18 was the major target of the kinase. We have now identified the sites on rat keratin 8 for cAMP-dependent protein kinase. Sequential analysis of the purified phosphoropeptides, together with the known primary sequence, revealed that four major sites, Ser-12, Ser-23, Ser-36, and Ser-50, and three minor sites, Ser-8, Ser-33, Ser-42, are located in the amino-terminal head domain, while three minor sites, Ser-416, Ser-423 and Ser-425 locate in the carboxyl-terminal tail domain.