Merck
BR114820
BRAND®PP刻度离心管,螺口盖
volume 50 mL, without base, non-sterile
别名:
离心管, 试管
登录并加购,请在购物车里查看协议价、货期和发货地。
选择规格
变更视图
关于此项目
NACRES:
NB.43
UNSPSC Code:
41121703
eCl@ss:
32040302
Material:
polypropylene , screw top cap, translucent tube
Sterility:
non-sterile
Packaging:
pack of 300
Feature:
RCF 3,000 × g, without base
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
material
polypropylene , screw top cap, translucent tube
sterility
non-sterile
feature
RCF 3,000 × g, without base
packaging
pack of 300
manufacturer/tradename
BRAND 114820
volume
50 mL
General description
The BRAND® PP Graduated Centrifuge Tube with Screw Cap is made from high-quality polypropylene (PP) and features a translucent design for easy visibility of contents. This tube is engineered to withstand relative centrifugal forces (RCF) of up to 3000, making it suitable for various laboratory applications. It comes graduated, allowing for precise measurements, and is available in both non-sterile and gamma-ray sterilized options in 15 ml (graduated up to 13 ml) or 50 ml capacities. The screw cap, made from polyethylene (PE) and equipped with a sealing cone, ensures a secure closure to prevent leaks. This centrifuge tube offers high clarity and reliability, making it an essential tool for researchers and technicians in the lab.
Features and Benefits
  • BRAND® graduated centrifuge tube is the ideal solution for all your centrifuge needs.
  • Made from highly transparent polypropylene (PP) for enhanced sample visibility.
  • Bio-inert PP offers optimal cell and protein recovery.
  • Includes a screw cap with a plug for preventing sample loss.
  • Tubes are tested to withstand RCF up to 3000.
  • γ-ray sterilized.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
历史批次信息供参考:
分析证书(COA)