Merck
BR122548
BRAND®实验室瓶子,有刻度和螺口盖
capacity 250 mL
登录查看公司和协议定价
别名:
实验室瓶子, 玻璃, 瓶子
UNSPSC代码:
41121800
NACRES:
NB.43
物料
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
接合处
Joints threaded neck (GL45)
无菌性
non-sterile
特点
autoclavable
graduations
with cap
包装
pack of 10 ea
制造商/商品名称
BRAND 122548
容量
250 mL
直径× 高度
70 mm × 144 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG