Merck
BR4600241
BRAND®Dispensette® S 固定容量瓶口分液器
volume 10 mL, with recirculation valve
登录查看公司和协议定价
NACRES:
NB.01
特点
DE-M marking
with recirculation valve
制造商/商品名称
BRAND 4600241
体积
10 mL
配件
(GL 28/S28)
GL 32
GL 38
S 40
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
BR4600240BR4600211BR4600210
manufacturer/tradename
BRAND 4600241
manufacturer/tradename
BRAND 4600240
manufacturer/tradename
BRAND 4600211
manufacturer/tradename
BRAND 4600210
feature
DE-M marking, with recirculation valve
feature
DE-M marking, without recirculation valve
feature
DE-M marking, with recirculation valve
feature
DE-M marking, without recirculation valve
fitting
GL 32, S 40, (GL 28/S28), GL 38
fitting
(GL 28/S28), GL 32, GL 38, S 40
fitting
(GL 28/S28), GL 32, GL 38, S 40
fitting
(GL 28/S28), GL 32, GL 38, S 40
一般描述
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
特点和优势
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
其他说明
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 1
1 of 1
相关内容
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.