Merck
BR759005
BRAND®大比色皿
polystyrene
别名:
比色皿
NACRES:
SB.32
物料
polystyrene
特点
minimum filling volume 2.5 mL
manufacturer/tradename
BRAND 759005
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
Show Differences
1 of 4
此商品
BR759105BR759035BR759030
BRAND® macro-cuvette polystyrene
BR759005
BRAND®大比色皿
BRAND® macro-cuvette PMMA
BR759105
BRAND®大比色皿
BRAND® macro-cuvette polystyrene, volume 2.5 mL, pack of 500
BR759035
BRAND®大比色皿
BRAND® macro-cuvette polystyrene, volume 2.5 mL, pack of 100
BR759030
BRAND®大比色皿
feature
minimum filling volume 2.5 mL
feature
minimum filling volume 2.5 mL
feature
four clear sided
feature
four clear sided
manufacturer/tradename
BRAND 759005
manufacturer/tradename
BRAND 759105
manufacturer/tradename
BRAND 759035
manufacturer/tradename
BRAND 759030
一般描述
  • BRAND® Macro cuvette is manufactured under controlled room conditions.
  • Ideal for the determinations in water analysis, chemistry, and life science applications.
  • Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Eliminates the risk of cross-contamination.
  • Grouped by mold cavity number.
  • An economical alternative to Quartz cuvettes.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 7
1 of 7
BRAND® UV cuvettes macro, volume 2.5 mL, pack of 100
BR759125
BRAND® UV比色皿
BRAND® UV cuvettes macro, volume 2.5 mL, pack of 500
BR759128
BRAND® UV比色皿
BRAND® semi-micro cuvette PMMA, minimum filling volume 1.5 mL
BR759115
BRAND®半微量比色皿
vibrant-m
Z652520
Greiner semi-micro cuvette
BRAND® UV cuvettes semi-micro, chamber volume 1.5-3 mL
Z628026
BRAND® 紫外比色皿
BRAND® cuvette rack Holds 16 cuvettes
BR759500
BRAND®比色皿架