Merck
DWK92812F-50
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (50 mL), class A
登录查看公司和协议定价
物料
PTFE stopper
glass flask
特点
class A
包装
case of 6 ea
manufacturer/tradename
Kimble® 92812F-50
培养瓶容量
(50 mL)
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
DWK92812F-10DWK92812F-200DWK92812F-250
material
PTFE stopper, glass flask
material
PTFE stopper, glass flask
material
PTFE stopper, glass flask
material
PTFE stopper, glass flask
manufacturer/tradename
Kimble® 92812F-50
manufacturer/tradename
Kimble® 92812F-10
manufacturer/tradename
Kimble® 92812F-200
manufacturer/tradename
Kimble® 92812F-250
packaging
case of 6 ea
packaging
case of 6 ea
packaging
case of 6 ea
packaging
case of 6 ea
flask capacity
(50 mL)
flask capacity
(10 mL)
flask capacity
(200 mL)
flask capacity
(250 mL)
一般描述
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
法律信息
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA