GF11639628

Sigma-Aldrich

foil, not light tested, 45x200mm, thickness 0.12mm, as rolled, 99.9%

别名:
线性分子式:
Li
CAS号:
分子量:
6.94
MDL number:
PubChem Substance ID:
价格与库存信息目前不能提供

assay

99.90%

form

foil

mfr. no.

Goodfellow, 116-396-28

resistivity

9.446 μΩ-cm, 20°C

L × W × thickness

45 mm × 200 mm × 0.12 mm

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

Li

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

正在寻找类似产品? Visit 产品对比指南

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

RIDADR

UN 1415 4.3 / PGI

Zhi Yang Tam et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 69(7), 810-820 (2014-01-09)
Aging has been associated with the accumulation of damages in molecules and organelles in cells, particularly mitochondria. The rate of damage accumulation is closely tied to the turnover of the affected cellular components. Perturbing mitochondrial turnover has been shown to...
Xiangbo Meng et al.
Advanced materials (Deerfield Beach, Fla.), 24(27), 3589-3615 (2012-06-16)
Lithium-ion batteries (LIBs) are used widely in today's consumer electronics and offer great potential for hybrid electric vehicles (HEVs), plug-in HEVs, pure EVs, and also in smart grids as future energy-storage devices. However, many challenges must be addressed before these...
Cary M Hayner et al.
Annual review of chemical and biomolecular engineering, 3, 445-471 (2012-04-25)
The lithium-ion battery is the most promising battery candidate to power battery-electric vehicles. For these vehicles to be competitive with those powered by conventional internal combustion engines, significant improvements in battery performance are needed, especially in the energy density and...
Jian Liu et al.
Chemical communications (Cambridge, England), 47(47), 12578-12591 (2011-08-25)
Yolk/shell or 'rattle-typed' nanomaterials with nanoparticle cores inside hollow shells are interesting among the complex hollow nanostructures. Yolk/shell nanoparticles (YSNs) are promising functional nanomaterials for a variety of applications such as catalysis, delivery, lithium-ion batteries and biosensors due to their...
Bellamkonda K Kishore et al.
American journal of physiology. Renal physiology, 304(9), F1139-F1149 (2013-02-15)
By virtue of its unique interactions with kidney cells, lithium became an important research tool in renal physiology and pathophysiology. Investigators have uncovered the intricate relationships of lithium with the vasopressin and aldosterone systems, and the membrane channels or transporters...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。