Merck
All Photos(2)

GF15420844

foil, thickness 0.03 mm, size 100 × 100 mm, purity 99.97%

Synonym(s):
BI000170
Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

蒸汽压

<0.1 mmHg ( 20 °C)

测定

99.97%

形式

foil

manufacturer/tradename

Goodfellow 154-208-44

电阻率

129 μΩ-cm, 20°C

尺寸

100 × 100 mm

厚度

0.03 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

密度

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

一般描述

For updated SDS information please visit www.goodfellow.com

包装

1, 2 ea

法律信息

Product of Goodfellow

储存分类代码

11 - Combustible Solids

WGK

nwg

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service