Merck
All Photos(2)

GF18049361

foil, thickness 0.7 mm, 15 mm diameter, purity 99%

Synonym(s):
NI000588
Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

测定

99%

形式

foil

manufacturer/tradename

Goodfellow 180-493-61

电阻率

6.97 μΩ-cm, 20°C

直径

15 mm
15 mm

厚度

0.7 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

密度

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

象形图

Exclamation markHealth hazard

警示用语:

Danger

危险声明

危险分类

Carc. 2 - Skin Sens. 1 - STOT RE 1

储存分类代码

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service