Merck
All Photos(1)
GF19411466
T1 - thermocouple alloy, Ni90Cr10
insulated wire, 20m length, 0.125mm conductor diameter, 0.015mm insulation thickness, polyester insulation
Synonym(s):
NI095840
NACRES:
NA.23
描述
polyester insulation
thermocouple wire
形式
wire (insulated)
manufacturer/tradename
Goodfellow 194-114-66
隔离层厚度
0.015 mm
电线长度 × 直径
20 m × 0.125 mm , conductor diameter
Looking for similar products? Visit Product Comparison Guide
一般描述
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
应用
may be used as one component of a thermocouple
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service