GF29507014
Molybdenum-rhenium alloy, Mo52.5Re47.5
200mm tube, 2mm outside diameter, 0.4mm wall thickness, 1.2mm inside diameter, as drawn
别名:
MO077201
价格与库存信息目前不能提供
form
tubes
mfr. no.
Goodfellow, 295-070-14
size L × wall thickness
200 mm × 0.4 mm
size O.D. × I.D.
2 mm × 1.2 mm
正在寻找类似产品? Visit 产品对比指南
General description
For updated SDS information please visit www.goodfellow.com.