Merck
All Photos(1)

GF31048635

Polyvinylchloride - Unplasticized

(UPVC), film, thickness 0.38 mm, L 1 m, clear

Synonym(s):
CV311450
NACRES:
NA.23

形式

film

manufacturer/tradename

Goodfellow 310-486-35

长度

1 m

厚度

0.38 mm

颜色

clear

Related Categories

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service