Merck
All Photos(1)

GF33528786

聚四氟乙烯

(PTFE), tube, outside diameter 0.37mm, L 1 m, wall thickness 0.15 mm

Synonym(s):
FP307003
NACRES:
NA.23

形式

tubes

manufacturer/tradename

Goodfellow 335-287-86

长度

1 m

壁厚度

0.15 mm

Related Categories

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service