Merck

GF50354692

foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%

别名:
Empirical Formula (Hill Notation):
Cr
CAS号:
分子量:
52.00
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

检测方案

99.99%

形式

foil

manufacturer/tradename

Goodfellow 503-546-92

电阻率

12.7 μΩ-cm, 20°C

尺寸 × 厚度

25 x 25 mm × 0.02 mm

bp

2672 °C (lit.)

mp

1857 °C (lit.)

密度

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

正在寻找类似产品? Visit 产品对比指南

一般描述

For updated SDS information please visit www.goodfellow.com.

法律信息

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

请输入1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

Chad M Thompson et al.
Critical reviews in toxicology, 43(3), 244-274 (2013-03-01)
Abstract Chronic exposure to high concentrations of hexavalent chromium (Cr(VI)) in drinking water causes intestinal adenomas and carcinomas in mice, but not in rats. Cr(VI) causes damage to intestinal villi and crypt hyperplasia in mice after only one week of
Yinan Hua et al.
The Journal of nutritional biochemistry, 23(4), 313-319 (2012-03-20)
Type 2 diabetes is often associated with obesity, dyslipidemia and cardiovascular anomalies and is a major health problem approaching global epidemic proportions. Insulin resistance, a prediabetic condition, precedes the onset of frank type 2 diabetes and offers potential avenues for
Satyendra Kumar Garg et al.
Reviews of environmental contamination and toxicology, 217, 75-140 (2012-02-22)
Bioremediation offers the possibility of using living organisms (bacteria, fungi, algae,or plants), but primarily microorganisms, to degrade or remove environmental contaminants, and transform them into nontoxic or less-toxic forms. The major advantages of bioremediation over conventional physicochemical and biological treatment
Yana Chervona et al.
Metallomics : integrated biometal science, 4(7), 619-627 (2012-04-05)
Carcinogenic metals, such as nickel, arsenic, and chromium, are widespread environmental and occupational pollutants. Chronic exposure to these metals has been connected with increased risks of numerous cancers and as well as non-carcinogenic health outcomes, including cardiovascular disease, neurologic deficits
Erik Wennerberg et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(22), 5733-5744 (2014-09-13)
Anaplastic thyroid carcinoma (ATC) is one of the most aggressive forms of cancer with no curative therapies available. To date, strategies to target ATC by immunotherapy have not been evaluated. We investigated whether ATC would be a suitable target for

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门