Merck
All Photos(1)
GF60966366
Sigma-Aldrich
rod, 70mm, diameter 10.0mm, cast sticks, 100%
Synonym(s):
Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23
蒸汽压
<0.1 mmHg ( 20 °C)
测定
100%
形式
rod
manufacturer/tradename
Goodfellow 609-663-66
电阻率
129 μΩ-cm, 20°C
长度 × 直径
70 mm × 10.0 mm
bp
1560 °C (lit.)
mp
271 °C (lit.)
密度
9.8 g/mL at 25 °C (lit.)
SMILES string
[Bi]
InChI
1S/Bi
InChI key
JCXGWMGPZLAOME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
一般描述
For updated SDS information please visit www.goodfellow.com.
包装
1, 2, 5, 10 ea
法律信息
Product of Goodfellow
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service