Merck
All Photos(2)

GF64636476

foil, thickness 0.25 mm, diameter 25 mm, purity 99.6+%

Synonym(s):
TI000410
Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

测定

≥99.6%

形式

foil

自燃温度

860 °F

manufacturer/tradename

Goodfellow 646-364-76

电阻率

42.0 μΩ-cm, 20°C

直径

25 mm

厚度

0.25 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

密度

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

储存分类代码

13 - Non Combustible Solids

WGK

nwg

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service