Merck
All Photos(2)

GF67024313

foil, thickness 0.020 mm, diameter 50 mm, purity 99%

Synonym(s):
AL000405
Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

测定

99%

形式

foil

自燃温度

1400 °F

manufacturer/tradename

Goodfellow 670-243-13

电阻率

2.6548 μΩ-cm

直径

50 mm

厚度

0.020 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

密度

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service