Merck
All Photos(2)
GF77615039
bolt, length 50 pcs, size m3 x 20 mm, head style hexagonal
Synonym(s):
宽度
Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL编号:
PubChem化学物质编号:
包装
pkg of 50
manufacturer/tradename
Goodfellow 776-150-39
电阻率
4.9 μΩ-cm, 20°C
bp
5660 °C (lit.)
mp
3410 °C (lit.)
密度
19.3 g/mL at 25 °C (lit.)
SMILES string
[W]
InChI
1S/W
InChI key
WFKWXMTUELFFGS-UHFFFAOYSA-N
一般描述
For updated SDS information please visit www.goodfellow.com
法律信息
Product of Goodfellow
储存分类代码
13 - Non Combustible Solids
WGK
nwg
闪点(F)
Not applicable
闪点(C)
Not applicable
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service