Merck

SIK4710-20

Sigma-Aldrich

11-Azidoundecyltrimethoxysilane

≥95%

登录to View Organizational & Contract Pricing

Select a Size

Empirical Formula (Hill Notation):
C14H31N3O3Si
分子量:
317.50
NACRES:
NA.23

检测方案

≥95%

形式

liquid (colorless)

保质期

3 years under the recommended conditions

应用

"CLICK CHEMISTRY"
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

法律信息

Product of SiKEMIA

象形图

Exclamation mark

警示用语:

Warning

危险声明

危险分类

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

靶器官

Respiratory system

储存分类代码

10 - Combustible liquids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

输入内容 1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

批号可以在产品标签上"批“ (Lot或Batch)字后面找到。

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

其他客户在看

Jeremiah C Traeger et al.
ACS nano, 13(7), 7850-7859 (2019-06-28)
Adsorption of soluble DNA to surfaces decorated with complementary DNA plays an important role in many bionanotechnology applications, and previous studies have reported complex dependencies of the surface density of immobilized DNA on hybridization. While these effects have been speculatively
Robert J Rawle et al.
Biophysical journal, 117(3), 445-452 (2019-07-22)
Fluorescent dye-dequenching assays provide a powerful and versatile means to monitor membrane fusion events. They have been used in bulk assays, for measuring single events in live cells, and for detailed analysis of fusion kinetics for liposomal, viral, and cellular
Jeremiah C Traeger et al.
Journal of colloid and interface science, 566, 369-374 (2020-02-06)
Single-molecule Förster Resonance Energy Transfer was used to observe the adsorption of fluorescently-labeled "target" DNA oligonucleotides and their association and hybridization with complementary DNA "probes" tethered to the surface as a function of surface grafting density. Ionic strength was varied

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门