Merck
WHA10441001
Whatman® glass vacuum filtration device
GV025 series, GV025/0/1 funnel, capacity 60 mL
别名:
Z750743
NACRES:
NB.24
物料
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
包装
pk of 1
manufacturer/tradename
Whatman 10441001
Whatman Article No. 28415541 (US reference)
容量
60 mL
适用性
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
Show Differences
1 of 4
此商品
WHA10443001WHA10442001WHA10442213
packaging
pk of 1
packaging
pk of 1
packaging
pack of 1
packaging
pk of 1
manufacturer/tradename
Whatman 10441001, Whatman Article No. 28415541 (US reference)
manufacturer/tradename
Whatman 10443001, Whatman Article No. 28415575 (US reference)
manufacturer/tradename
Whatman 10442001, Whatman Article No. 28415552 (US reference)
manufacturer/tradename
Whatman 10442213, Whatman Article No. 28415567 (US reference)
capacity
60 mL
capacity
-
capacity
-
capacity
-
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
一般描述
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
其他说明
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
法律信息
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA