WHA10442203
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
pk of 1
Whatman 10442203
Whatman Article No. 28415565 (US reference)
1000 mL
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
正在寻找类似产品? Visit 产品对比指南
1 of 4
此商品 | WHA10442213 | WHA10441203 | WHA10441001 |
---|---|---|---|
packaging pk of 1 | packaging pk of 1 | packaging pk of 1 | packaging pk of 1 |
manufacturer/tradename Whatman 10442203, Whatman Article No. 28415565 (US reference) | manufacturer/tradename Whatman 10442213, Whatman Article No. 28415567 (US reference) | manufacturer/tradename Whatman 10441203, Whatman Article No. 28415550 (US reference) | manufacturer/tradename Whatman 10441001, Whatman Article No. 28415541 (US reference) |
bottle capacity 1000 mL | bottle capacity 1000 mL | bottle capacity - | bottle capacity - |
neck diam.
| neck diam. - | neck diam. - | neck diam. - |
suitability suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions) | suitability suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions) | suitability suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions) | suitability suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions) |
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
如何查找产品货号
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
如何查找COA批号
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。