Merck
WHAWB120411
QIAcard FTA Elute Micro cards
Indicating, one sample area per card, pkg of 100 cards
登录查看公司和协议定价
别名:
Z747572
NACRES:
NB.22
manufacturer/tradename
Qiagen WB120411
包装
pkg of 100 cards
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
WHAWB120410WHAWB120401WHAWB120310
packaging
pkg of 100 cards
packaging
pkg of 100 cards
packaging
pkg of 25 cards
packaging
pkg of 25 cards
一般描述
FTA Cards offer a compact room-temperature storage system that reduces the need for precious freezer space. Unlike the QIAcard FTA cards, the FTA Elute cards feature a technology for convenient water based elution of nucleic acids.
QIAcard FTA Elute Micro format with one spot area (spot volume 12-40 µL) to collect, stabilize, process, transport, and archive colored samples such as blood. FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Convenient for protocols that require different locations for testing and archiving samples. Different samples can be processed independently.
特点和优势
  • Includes a pink dye that turns white when a colorless sample is applied.
  • One sample area for application of up to 40 μl sample volume per card.
  • Simply apply your sample to the FTA Card. Cell membranes and organelles are lysed and the released nucleic acids are entrapped in the fibers of the matrix. The nucleic acids remain immobilized and are preserved for transport, immediate processing or long-term room temperature storage.
  • Since captured nucleic acids are preserved, FTA Cards facilitate sample collection in remote locations and simplify sample transport.
  • Ship your samples back to the laboratory without expensive special handling or dry ice and process at your convenience.
  • FTA Cards can be used with virtually any sample type: Blood, Cultured cells, Buccal cells, Plasmids, and Solid tissue.
  • Captured nucleic acids are ready for purification when you are. The FTA Elute technology enables convenient water-based elution of nucleic acids for downstream experiments.
其他说明
Field of Use : For internal research use only
法律信息
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 10
1 of 10
QIAcard™ Indicating FTA™ Cards classic, 4 sample areas per card, pkg of 100 cards
WHAWB120206
QIAcard Indicating FTA Cards
QIAcard™ FTA™ Wash Buffer Nucleic acid purification buffer, bottle of 500 mL
WHAWB120204
QIAcard FTA Wash Buffer
QIAcard™ Indicating FTA™ Cards classic, 4 sample areas per card, pkg of 25 cards
WHAWB120306
QIAcard Indicating FTA Cards
Qiagen® Multi Barrier Pouch medium, size , L x W 4" x 4.5" (10.1 cm x 11.4 cm), pack of 100 ea
WHAWB100092
Qiagen® Multi Barrier Pouch
EasiCollect (50) device for collection of buccal DNA samples, pkg of pack of 50 devices
WHAWB120462
EasiCollect (50)
QIAcard™ Indicating FTA™ Cards mini, 2 sample areas per card, pkg of 100 cards
WHAWB120056
QIAcard Indicating FTA Cards
QIAcard™ FTA™ Gene cards 3 sample areas per card, pkg of 100 cards
WHAWB120208
QIAcard FTA Gene cards
Whatman® protein saver cards 903 Protein saver snap apart card, pkg of 100 ea
WHA10534320
Whatman® protein saver cards
商品
This page shows downstream applications after genomic DNA purification.
Description of common problems that can occur during genomic DNA preparation
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
实验方案
This page shows genomic DNA purification from sample on FTA elute from Cytiva.
A number of options are available for sample collection, transport, archiving, and DNA purification.
Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.