Merck
Z146978
Sigma-Aldrich
Lecture-bottle control valve
Monel®, hose outlet I.D. 1/4 in.
登录查看公司和协议定价
别名:
Monel® lecture-bottle control valve
NACRES:
NB.51
物料
Monel®
接合处
joint CGA Inlet
joint hose barb
参数
-65-250 °F temperature (-54-120 °C)
3,000 psi pressure range (@ 70°F)
软管出口内径
1/4 in.
配件
CGA 110 male
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z187224Z146951Z176052
Lecture-bottle control valve Monel®, hose outlet I.D. 1/4 in.
Z146978
Lecture-bottle control valve
Lecture-bottle control valve stainless steel, hose outlet I.D. 1/4 in.
Z146951
Lecture-bottle control valve
Lecture-bottle valve CGA Inlet for 180M/110F, 316 stainless steel
Z176052
Lecture-bottle valve
fitting
CGA 110 male
fitting
-
fitting
CGA 110 male
fitting
CGA Inlet for 180M/110F
parameter
-65-250 °F temperature (-54-120 °C), 3,000 psi pressure range (@ 70°F)
parameter
-
parameter
-65-250 °F temperature (-54-120 °C), 3,000 psi pressure range (@ 70°F)
parameter
-
material
Monel®
material
stainless steel
material
stainless steel
material
316 stainless steel
hose outlet I.D.
1/4 in.
hose outlet I.D.
-
hose outlet I.D.
1/4 in.
hose outlet I.D.
-
一般描述
CGA110, straight-pattern valve for precise flow control of lecture-bottle gas. Requires CGA110 PTFE sealing washer.
Brass control valve shown.
警告
Gas-control valves do not control pressure in a closed system. Regulator or relief valves should be used to prevent the buildup of unsafe pressures that could lead to explosion.
法律信息
Monel is a registered trademark of Huntington Alloys Corp.
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 4
1 of 4
"T"-purge valve CGA Inlet size 180
Sigma-Aldrich
Z151661
"T"-purge valve
阀门 stainless steel diaphragm, for CGA 110/180 right angle path, internal/external, right hand thread
Sigma-Aldrich
Y906611
阀门
Sigma-Aldrich
Sigma-Aldrich
Z146994
PTFE sealing washers