Merck
Z165344
Large laboratory assortment of black-rubber stoppers
pkg of approx. 66 stoppers
登录to View Organizational & Contract Pricing
Select a Size
别名:
rubber stopper, stopper, stoppers
NACRES:
NB.35
包装
pkg of approx. 66 stoppers
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z165204Z166065Z764612
Red-rubber micro stoppers L 50 mm
Z165204
Red-rubber micro stoppers
Rubber Stoppers gray butyl rubber, size 20 mm
Supelco
Z166065
橡胶塞
一般描述
Approx. 66 stoppers, including solid, 1 and 2 hole, in sizes 00 to 8. 2lb package.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA