Z173231
1 of 10
Sigma-Aldrich
Z173126
Reaction and storage tube with glass stopcock on side-arm
Z164046
Aldrich® cold-finger condenser
Z515981
Schlenk reaction and storage tube
Z173061
Solvent storage flask with glass stopcock on side-arm
Z409235
Aldrich® Schlaker reaction tube
Z225789
Aldrich® cold-finger for high-efficiency cold-trap
SYNF891410
Synthware™ reaction tube with glass stopcock
Z166898
Aldrich® 气敏化学玻璃实验室套件
SYNC341014
Synthware™ Dewar microscale condenser
Z164038
borosilicate glass round bottom two-neck
R
female joint: ST/NS14/20
non-sterile
closure type female Joint: ST/NS
pack of 1
25 mL
正在寻找类似产品? 访问 产品对比指南
玻璃耗材
批号
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 9
Z167517
Aldrich® Schlenk-type Vigreux distilling column
Z515698
Schlenk Kjeldahl reaction and storage flask
Z220280
Reaction flask with glass stopcock on side-arm
Z220140
Filter funnel with drip-tip and two side-arms each equipped with a glass stopcock
Z173363
Recrystallizer
Z220434
Sealing tube
SYNF901425
Synthware™ pear-shaped Schlenk flask with stopcock
Z173088
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.