Merck
Z510785
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 14/20, thread size 13-425
螺纹尺寸
13-425
接合处
female joint: ST/NS14/20
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
Show Differences
1 of 4
此商品
Z510815Z510793Z510807
Connecting Adapter female joint: ST/NS 14/20, thread size 13-425
Sigma-Aldrich
Z510785
Connecting Adapter
Connecting Adapter female joint: ST/NS 14/20, thread size 20-400
Sigma-Aldrich
Z510815
Connecting Adapter
Connecting Adapter female joint: ST/NS 19/22, thread size 13-425
Sigma-Aldrich
Z510793
Connecting Adapter
Connecting Adapter female joint: ST/NS 24/40, thread size 13-425
Sigma-Aldrich
Z510807
Connecting Adapter
thread size
13-425
thread size
20-400
thread size
13-425
thread size
13-425
一般描述
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 9
1 of 9