Merck
All Photos(2)
Z527645
塑料桶塞和盖
NPT threaded plug for metal drums, Rieke 2 inch, with micro-porous vent hole
NACRES:
NB.51
包装
pkg of 5 ea
Looking for similar products? Visit Product Comparison Guide
一般描述
Rieke®的插头和盖子是带有 EPDM 垫圈的 HDPE。Tri-Sure 插头是带有橡胶垫圈的 PP。符合 FDA 和 UN 包装标准。鼓形水龙头可安装 3/4 英寸的插头。NPT 中心孔。
法律信息
Rieke is a registered trademark of Rieke Metals, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service