Merck
All Photos(1)
Z530174
Sigma-Aldrich
Manual gas control valve
stainless steel, CGA Inlet size 240, hose barb outlet
NACRES:
NB.51
物料
stainless steel
inlet pressure measuring range
≤3,000 psig
CGA入口尺寸
240
Looking for similar products? Visit Product Comparison Guide
一般描述
Provides an economical means of controlling gas flow directly from the cylinder. Manufactured with SS or Monel® bodies to accommodate corrosive gases that cannot be run through a regulator. Valve stem packing is PTFE. Hose barb outlet.
警告
Control valves do not regulate pressure, they only control flow.
法律信息
Monel is a registered trademark of Huntington Alloys Corp.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service