Z530883
Sigma-Aldrich
Flexible PTFE adapter
top female Joint: ST/NS 24/40, bottom male Joint: ST/NS 19/22
价格与库存信息目前不能提供
Joint
female Joint: ST/NS top24/40
male Joint: ST/NS bottom19/22
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Used for the transfer of powders and liquids. Can be compressed to approximately 190 mm or expanded to 250 mm. Glass joints with flexible corrugated PTFE tubing is pliable enough to create an offset adapter.
Other Notes
Picture is an example of male and female joint style.