Merck
All Photos(1)
Z531197
Sigma-Aldrich
Aldrich® coiled condenser
unjacketed, L 500 mm, joint: ST/NS 29/32
NACRES:
NB.43
接合处
joint: ST/NS29/32
长度
500 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
一般描述
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service