Z531804
Sigma-Aldrich
Tubing Adapters with Torion Connection
With hose barb, O.D. 1/2  in., male Joint: ST/NS 29/32
价格与库存信息目前不能提供
Joint
male Joint: ST/NS29/32
O.D.
1/2  in.
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
The unique Torion connection ensures an airtight seal between tubing and reaction vessels when transferring air-sensitive materials.
5/16 in. O.D fits a #8 Torion, 1/2 in. O.D. fits a #14 Torion, and 3/16 in. O.D fits a #5 Torion.
  • Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE-coated sealing ring of the adapter
  • Turn the bushing to make an airtight compression seal with the tubing
  • Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use
  • O.D. refers to the tubing outer diameter
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.