Z543527
Best® N-DEX® free disposable nitrile gloves
textured tips, size XL
NACRES:
NB.35
价格与库存信息目前不能提供
material
green
nitrile
size
XL
sterility
non-sterile
feature
disposable
latex free
powder-free
packaging
pack of 100 ea (boxes of 100)
mfr. no.
Best Manufacturing, 7705PFTXL
L × thickness
9.5 in. × 4 mil
color
green
正在寻找类似产品? Visit 产品对比指南
General description
Developed by Best to the same exacting standards of its N-DEX family of hand protection.
N-DEX Free′s accelerator-free formulation creates a luminous, fun, yet PPE-conscious, high-visibility green glove.
  • Powder-free, ambidextrous
  • Free from accelerators common in all disposable nitrile gloves, which have been linked to workplace conditions like Type IV dermatitis
  • Free from natural rubber latex and Type I allergic reactions linked to latex proteins
Legal Information
Best is a registered trademark of Showa Glove Co.
N-DEX is a registered trademark of Showa Glove Co.